Sodium-2-ethylhexanoate ≥97%
Supplier: STREM CHEMICALS INC MS
Synonyms:
2-Ethylcaproic acid sodium salt, Sodium 2-ethylhexanoate, 2-Ethylhexanoic acid sodium salt
93-1163-100G
93-1163-25G
100205-282EA
352.43
USD
100205-282
100202-698
Sodium-2-ethylhexanoate ≥97%
Sodium-2-ethylhexanoate
Formula:
C₈H₁₅NaO₂ MW: 166.2 g/mol |
MDL Number:
MFCD00014007 CAS Number: 19766-89-3 EINECS: 243-283-8 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...