Sodium hexafluorostannate(IV) ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-1148-25G
93-1148-5G
100204-150EA
99.08
USD
100204-150
100205-278
Sodium hexafluorostannate(IV) ≥99%
Sodium hexafluorostannate(IV)
Formula:
F₆Na₂Sn MW: 278.68 g/mol |
MDL Number:
MFCD00058776 CAS Number: 16924-51-9 EINECS: 240-988-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...