Sodium hexafluorosilicate ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

93-1147-100G 93-1147-500G
100205-276EA 42.46 USD
100205-276 100204-148
Sodium hexafluorosilicate ≥99%
Sodium hexafluorosilicate
Formula: F₆Na₂Si
MW: 188.06 g/mol
Melting Pt: Dec.
Density: 2.68 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003491
CAS Number: 16893-85-9
EINECS: 240-934-8
UN: 2674
ADR: 6.1,III
Merck Index: 12,08769

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR