Sodium chlorate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

93-1126-1000G 93-1126-5000G
100196-454EA 65.11 USD
100196-454 100210-156
Sodium chlorate ≥99%
Sodium chlorate
Formula: ClNaO₃
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 248…261 °C (dec.)
Density: 2.490
Storage Temperature: Ambient
MDL Number: MFCD00003479
CAS Number: 7775-09-9
EINECS: 231-887-4
UN: 1495
ADR: 5.1,II
REACH: 01-2119474389-23
Merck Index: 13,08670

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR