Sodium carbonate, anhydrous ≥99.5% ACS

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Calcined soda, Carbonic acid disodium salt

93-1125-2000G 93-1125-500G
100205-272EA 280.25 USD
100205-272 100205-274
Sodium carbonate, anhydrous ≥99.5% ACS
Sodium carbonate
Formula: Na₂CO₃
MW: 105.99 g/mol
Boiling Pt: 1600
Melting Pt: 854
Density: 2.532
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR