Sodium borohydride ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Sodium tetrahydridoborate, Sodium tetrahydroborate

93-1118-50G 93-1118-250G
100197-666EA 62.28 USD
100197-666 100201-058
Sodium borohydride ≥98%
Sodium borohydride
Formula: NaBH₄
Melting Pt: ~400 °C (dec.)
Density: 1.07 g/cm³ (20 °C)
Flash Pt: 69 °C
Storage Temperature: Ambient
MDL Number: MFCD00003518
CAS Number: 16940-66-2
EINECS: 241-004-4
UN: 1426
ADR: 4.3,I
Merck Index: 13,08664

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR