di-Sodium tetraborate, anhydrous ≥99.998% (B basis) (50 ppm K), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

93-1072-50G 93-1072-10G
100210-154EA 447.26 USD
100210-154 100197-658
di-Sodium tetraborate, anhydrous ≥99.998% (B basis) (50 ppm K), PURATREM
di-Sodium tetraborate
Formula: Na₂B₄O₇
Boiling Pt: 1575 °C (1013 hPa)
Melting Pt: 742 °C
Density: 2.38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00081185
CAS Number: 1330-43-4
EINECS: 215-540-4
REACH: 01-2119490790-32
Merck Index: 12,08733

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR