di-Sodium tetraborate decahydrate ≥99.5% ACS

Supplier: STREM CHEMICALS INC MS
Danger

93-1070-500G 93-1070-2000G
100204-136EA 67.94 USD
100204-136 100198-288
di-Sodium tetraborate decahydrate ≥99.5% ACS
di-Sodium tetraborate decahydrate
Formula: Na₂B₄O₇·10H₂O
Boiling Pt: 320 °C (1013 hPa)
Melting Pt: 75 °C (dec.)
Density: 1.73 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149193
CAS Number: 1303-96-4
EINECS: 215-540-4
REACH: 01-2119490790-32
Merck Index: 13,08662

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR