Sodium hydrogensulfide, anhydrous
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Sodium hydrogen sulphide
93-1061-1G
93-1061-5G
93-1061-25G
100201-844EA
97.66
USD
100201-844
100201-848
100201-846
Sodium hydrogensulfide, anhydrous
Sodium hydrogensulphide
Formula:
HNaS MW: 56.06 g/mol Melting Pt: 350 °C Density: 1.79 g/cm³ (20 °C) |
MDL Number:
MFCD00011124 CAS Number: 16721-80-5 EINECS: 240-778-0 UN: 2318 ADR: 4.2,II Merck Index: 12,08730 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...