Sodium hydrogen carbonate ≥99.7% ACS
Supplier: STREM CHEMICALS INC MS
Synonyms:
Sodium bicarbonate
93-1053-2000G
93-1053-500G
100201-840EA
142.95
USD
100201-840
100201-842
Sodium hydrogen carbonate ≥99.7% ACS
Sodium hydrogen carbonate
Formula:
NaHCO₃ MW: 84.01 g/mol Melting Pt: 270 °C Density: 2.159 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003528 CAS Number: 144-55-8 EINECS: 205-633-8 Merck Index: 13,08655 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...