Sodium hexafluorotitanate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Sodium hexafluorotitanate(IV)
93-1007-10G
93-1007-50G
100197-652EA
36.8
USD
100197-652
100201-834
Sodium hexafluorotitanate ≥98%
Sodium hexafluorotitanate
Formula:
F₆Na₂Ti MW: 207.85 g/mol |
MDL Number:
MFCD00014244 CAS Number: 17116-13-1 EINECS: 241-181-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...