Nitrosium tetrafluoroborate ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Nitrosyl tetrafluoroborate, Nitrosonium tetrafluoroborate

93-0724-25G 93-0724-5G
100195-926EA 383.57 USD
100195-926 100210-146
Nitrosium tetrafluoroborate ≥97%
Nitrosium tetrafluoroborate
Formula: BF₄NO
MW: 116.81 g/mol
Boiling Pt: 250 °C (0.01 mmHg) (subl.)
Density: 2.185
MDL Number: MFCD00011433
CAS Number: 14635-75-7
EINECS: 238-679-2
UN: 1759
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR