Carbon ≥99.999%, rods

Supplier: STREM CHEMICALS INC MS
Warning

93-0605-1ROD 93-0605-5ROD
100210-136EA 35.38 USD
100210-136 100210-138
Carbon ≥99.999%, rods
Carbon
Formula: C
Boiling Pt: 4200 °C
Melting Pt: 3652…3697 °C (subl.)
Density: 3.65 g/cm³ (20 °C)
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
Merck Index: 13,01818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR