Phenylboronic acid ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-0570-10G
93-0570-50G
100196-438EA
79.26
USD
100196-438
100208-078
Phenylboronic acid ≥97%
Phenylboronic acid
Boronic Acid and Ester
Formula:
C₆H₇BO₂ Melting Pt: 215 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002103 CAS Number: 98-80-6 EINECS: 202-701-9 Merck Index: 13,01068 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...