Orthoboric acid ≥99.99% (B basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid

93-0559-1000G 93-0559-250G
100196-436EA 114.65 USD
100196-436 100208-492
Orthoboric acid ≥99.99% (B basis), PURATREM
Orthoboric acid
Formula: B(OH)₃
MW: 61.83 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 160 °C
Density: 1.44 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
REACH: 01-2119486683-25
Merck Index: 13,01326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR