Diboron trioxide ≥99.6% (B basis)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Boron oxide, Boric anhydride

93-0526-250G 93-0526-1000G
100197-640EA 35.38 USD
100197-640 100201-820
Diboron trioxide ≥99.6% (B basis)
Diboron trioxide
Formula: B₂O₃
MW: 69.62 g/mol
Boiling Pt: ~1860 °C
Melting Pt: 450 °C
Density: 1.81 to 1.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011315
CAS Number: 1303-86-2
EINECS: 215-125-8
REACH: 01-2119486655-24
Merck Index: 13,01327

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR