Boron tribromide ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

93-0514-100G 93-0514-25G 93-0514-500G
100207-758EA 161.35 USD
100207-758 100206-892 100202-434
Boron tribromide ≥99%
Boron tribromide

Halides for CVD & ALD, Volatile Precursors for CVD

Formula: BBr₃
Boiling Pt: 91.3 °C
Melting Pt: –46 °C
Density: 2.645 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011312
CAS Number: 10294-33-4
EINECS: 233-657-9
UN: 2692
ADR: 8,I
Merck Index: 13,01337

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR