Ammonium tetrafluoroborate ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Ammonium fluoroborate, ammonium fluoborate ammonium borofluoride ammonium fluoroborate [qr] ammonium tetrafluoroborate

93-0501-250G 93-0501-1000G
100210-128EA 124.55 USD
100210-128 100198-936
Ammonium tetrafluoroborate ≥99%
Ammonium tetrafluoroborate
Formula: H₄BF₄N
MW: 104.84
Melting Pt: 510 (subl.)
Density: 1.871
MDL Number: MFCD06245785
CAS Number: 13826-83-0
EINECS: 237-531-4
UN: 1759
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR