Lithium aluminum hydride ≥95%, powder
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Aluminium lithium hydride, Lithium tetrahydridoaluminate
93-0393-10G
93-0393-50G
100210-126EA
90.58
USD
100210-126
100206-888
Lithium aluminum hydride ≥95%, powder
Lithium aluminum hydride
Formula:
H₄AlLi MW: 37.95 g/mol Melting Pt: 125 °C (dec.) Density: 0.91 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011075 CAS Number: 16853-85-3 EINECS: 240-877-9 UN: 1410 ADR: 4.3,I Merck Index: 13,00348 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...