Lithium iodide, anhydrous ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

93-0382-100G 93-0382-25G
100210-754EA 458.58 USD
100210-754 100206-886
Lithium iodide, anhydrous ≥98%
Lithium iodide
Formula: ILi
MW: 133.85 g/mol
Boiling Pt: 1177 °C (1013 hPa)
Melting Pt: 450 °C
Density: 3.49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011092
CAS Number: 10377-51-2
EINECS: 233-822-5
Merck Index: 13,05557

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR