Lithium sulfide ≥98% (99.9%-Li)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-0378-5G
93-0378-25G
100205-248EA
147.2
USD
100205-248
100208-704
Lithium sulfide ≥98% (99.9%-Li)
Lithium sulphide
Formula:
Li₂S Boiling Pt: 1527 °C (1013 hPa) Melting Pt: > 900 °C Density: 1.66 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00011085 CAS Number: 12136-58-2 EINECS: 235-228-1 UN: 3178 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...