Lithium deuteride ≥99% (Isotopic)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Lithium hydride-D1
93-0374-1G
93-0374-5G
100205-246EA
433.11
USD
100205-246
100198-044
Lithium deuteride ≥99% (Isotopic)
Lithium deuteride
Formula:
LiD MW: 8.94 Melting Pt: 680 Density: 0.820 Storage Temperature: Ambient |
MDL Number:
MFCD00011091 CAS Number: 13587-16-1 EINECS: 237-018-5 UN: 1414 ADR: 4.3 I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...