Lithium metaborate dihydrate
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-0370-500G
93-0370-100G
100203-346EA
326.95
USD
100203-346
100198-932
Lithium metaborate dihydrate
Lithium metaborate dihydrate
Formula:
BLiO₂·2H₂O Density: 1,8 |
MDL Number:
MFCD00149768 CAS Number: 15293-74-0 EINECS: 236-631-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...