Lithium ≥99.8%

Supplier: STREM CHEMICALS INC MS
Danger

93-0369-25G 93-0369-100G
100204-122EA 292.98 USD
100204-122 100197-188
Lithium ≥99.8%
Lithium
Formula: Li
Boiling Pt: 1317 °C
Melting Pt: 179 °C
Density: 0.534
Storage Temperature: Ambient
MDL Number: MFCD00134051
CAS Number: 7439-93-2
EINECS: 231-102-5
UN: 1415
ADR: 4.3,I
Merck Index: 13,05542

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR