di-Lithium tetraborate ≥99.998% (Li basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Lithium tetraborate, Dilithium tetraborate
93-0365-10G
93-0365-50G
100201-810EA
70.77
USD
100201-810
100210-846
di-Lithium tetraborate ≥99.998% (Li basis), PURATREM
di-Lithium tetraborate
Formula:
Li₂B₄O₇ MW: 169.12 g/mol Melting Pt: 917 °C Density: 1.4 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011083 CAS Number: 12007-60-2 EINECS: 234-514-3 Merck Index: 14,05524 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...