Lithium wolframate ≥98%
Supplier: STREM CHEMICALS INC MS
Synonyms:
Lithium tungsten oxide, Lithium tungstate
93-0357-250G
93-0357-50G
100207-270EA
655.32
USD
100207-270
100203-344
Lithium wolframate ≥98%
Lithium wolframate
Formula:
Li₂O₄W MW: 261.72 g/mol Melting Pt: 742 °C Density: 3.71 |
MDL Number:
MFCD00016185 CAS Number: 13568-45-1 EINECS: 236-978-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...