Lithium tetrafluoroborate ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

93-0356-50G 93-0356-10G
100210-676EA 274.58 USD
100210-676 100200-020
Lithium tetrafluoroborate ≥98%
Lithium tetrafluoroborate
Formula: BF₄Li
Melting Pt: Dec.
Density: 0.852 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011087
CAS Number: 14283-07-9
EINECS: 238-178-9
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR