Lithium perchlorate, anhydrous ≥95% ACS
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-0345-50G
93-0345-250G
100201-808EA
49.54
USD
100201-808
100207-752
Lithium perchlorate, anhydrous ≥95% ACS
Lithium perchlorate
Formula:
ClLiO₄ MW: 106.39 g/mol Boiling Pt: 430 °C Melting Pt: 235 °C Density: 2.42 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011079 CAS Number: 7791-03-9 EINECS: 232-237-2 UN: 1481 ADR: 5.1,II Merck Index: 13,05561 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...