Lithium-2-ethylhexanoate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

93-0341-25G 93-0341-5G
100201-804EA 239.2 USD
100201-804 100201-806
Lithium-2-ethylhexanoate ≥98%
Lithium-2-ethylhexanoate
Formula: C₈H₁₅LiO₂
MDL Number: MFCD00058702
CAS Number: 15590-62-2
EINECS: 239-657-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR