Trilithium nitride ≥99.5% (Li basis)

Supplier: STREM CHEMICALS INC MS
Danger

93-0340-25G 93-0340-5G
100210-844EA 220.8 USD
100210-844 100206-880
Trilithium nitride ≥99.5% (Li basis)
tri-Lithium nitride
Formula: Li₃N
MW: 34.83 g/mol
Melting Pt: 845 °C
Density: 1.38 g/cm³ (20 °C)
MDL Number: MFCD00016186
CAS Number: 26134-62-3
EINECS: 247-475-2
UN: 2806
ADR: 4.3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR