Lithium niobate ≥99.998% (Nb basis), PURATREM

Supplier: STREM CHEMICALS INC MS

Synonyms: Lithium niobium oxide

93-0338-100G 93-0338-25G
100210-120EA 546.34 USD
100210-120 100202-516
Lithium niobate ≥99.998% (Nb basis), PURATREM
Lithium niobate
Formula: LiNbO₃
MW: 147.85
Melting Pt: 1257
Density: 4.638 g/cm³ (20 °C)
MDL Number: MFCD00011080
CAS Number: 12031-63-9
EINECS: 234-755-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR