Lithium methoxide ≥95%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Lithium methanolate, Lithium methylate
93-0336-50G
93-0336-250G
100203-830EA
179.75
USD
100203-830
100208-482
Lithium methoxide ≥95%
Lithium methoxide
Alkoxide
Formula:
CH₃LiO MW: 37.98 g/mol Boiling Pt: 64.6 °C (1013 hPa) Melting Pt: 500 °C Density: 0.865 g/cm³ (20 °C) |
MDL Number:
MFCD00036357 CAS Number: 865-34-9 EINECS: 212-737-7 UN: 3206 ADR: 4.2,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...