Lithium hydroxide monohydrate ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

93-0329-2000G 93-0329-500G
100203-342EA 2145.72 USD
100203-342 100198-928
Lithium hydroxide monohydrate ≥98%
Lithium hydroxide monohydrate
Formula: LiOH·1H₂O
MW: 41.96 g/mol
Boiling Pt: 920 °C (1013 hPa)
Melting Pt: 462 °C
Density: 1.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149772
CAS Number: 1310-66-3
EINECS: 215-183-4
UN: 2680
ADR: 8,II
Merck Index: 13,05556

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR