Lithium hydride ≥95%

Supplier: STREM CHEMICALS INC MS
Danger

93-0328-50G 93-0328-250G
100209-402EA 151.45 USD
100209-402 100209-400
Lithium hydride ≥95%
Lithium hydride
Formula: HLi
MW: 7.95 g/mol
Melting Pt: 680 °C
Density: 0.685 to 0.725 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011074
CAS Number: 7580-67-8
EINECS: 231-484-3
UN: 1414
ADR: 4.3,I
Merck Index: 13,05555

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR