Lithium fluoride ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-0322-250G
93-0322-1000G
100206-876EA
328.37
USD
100206-876
100197-186
Lithium fluoride ≥99%
Lithium fluoride
Formula:
FLi MW: 25.94 g/mol Boiling Pt: 1672 °C Melting Pt: 842 °C Density: 2.64 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011090 CAS Number: 7789-24-4 EINECS: 232-152-0 UN: 3288 ADR: 6.1,III Merck Index: 13,05553 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...