Lithium fluoride ≥99.9% (99.9+%-Li)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-0321-50G
93-0321-10G
93-0321-250G
100202-782EA
328.37
USD
100202-782
100208-996
100199-266
Lithium fluoride ≥99.9% (99.9+%-Li)
Lithium fluoride
Formula:
FLi MW: 25.94 g/mol Boiling Pt: 1676 Melting Pt: 870 Density: 2.635 Storage Temperature: Ambient |
MDL Number:
MFCD00011090 CAS Number: 7789-24-4 EINECS: 232-152-0 UN: 3288 ADR: 6.1 III Merck Index: 13,05553 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...