Lithium benzoate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Benzoic acid lithium salt

93-0308-100G 93-0308-500G
100205-232EA 55.2 USD
100205-232 100204-662
Lithium benzoate ≥99%
Lithium benzoate
Formula: C₇H₅LiO₂
MW: 128.06
Melting Pt: 300
Density: 1.397 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00035540
CAS Number: 553-54-8
EINECS: 209-042-6
Merck Index: 12,05547

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR