Triammonium dodecamolybdophosphate hydrate, Reagent Grade

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Triammonium 12-molybdophosphate hydrate, Phosphomolybdic acid, ammonium salt hydrate, Ammonium phosphomolybdate hydrate

93-0248-50G 93-0248-250G
100198-926EA 49.54 USD
100198-926 100205-228
Triammonium dodecamolybdophosphate hydrate, Reagent Grade
Triammonium dodecamolybdophosphate hydrate
Formula: H₁₂Mo₁₂N₃O₄₀P·H₂O
MW: 1894.36 g/mol
Melting Pt: Dec.
MDL Number: MFCD00150423
CAS Number: 54723-94-3
EINECS: 234-714-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR