Diammonium hydrogen orthophosphate ≥98% ACS

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Diammonium hydrogen orthophosphate, Ammonium hydrogen phosphate, di-Ammonium hydrogen phosphate, Ammonium Phosphate, Ammonium biphosphate, Fyrex, Ammonium phosphate dibasic, Ammonium monohydrogen phosphate, Diammonium hydrogen phosphate

93-0220-1000G 93-0220-250G
100196-416EA 232.12 USD
100196-416 100201-790
Diammonium hydrogen orthophosphate ≥98% ACS
Diammonium hydrogen orthophosphate
Formula: (NH₄)₂HPO₄
MW: 132.06 g/mol
Melting Pt: 185
Density: 1.619 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010891
CAS Number: 7783-28-0
EINECS: 231-987-8
REACH: 01-2119490974-22
Merck Index: 13,00546

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR