Ammonium hydrogen difluoride ≥96% (contains 3-4% NH4F)

Supplier: STREM CHEMICALS INC MS
Danger

93-0205-500G 93-0205-2000G
100196-412EA 63.69 USD
100196-412 100199-460
Ammonium hydrogen difluoride ≥96% (contains 3-4% NH4F)
Ammonium hydrogen difluoride
Formula: NH₄HF₂
MW: 57.04 g/mol
Melting Pt: 125.6 °C
Density: 0.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012018
CAS Number: 1341-49-7
EINECS: 215-676-4
UN: 1727
ADR: 8,II
Merck Index: 13,00498

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR