Ammonium acetate, anhydrous ≥97% ACS
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-0201-2000G
93-0201-500G
100197-178EA
460
USD
100197-178
100197-180
Ammonium acetate, anhydrous ≥97% ACS
Ammonium acetate
Formula:
H₃CCOONH₄ Melting Pt: 114 °C Density: 1.17 g/cm³ (20 °C) Flash Pt: 136 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013066 CAS Number: 631-61-8 EINECS: 211-162-9 Merck Index: 13,00495 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...