Bismuth ≥99.999%, shot, elongated

Supplier: STREM CHEMICALS INC MS

83-0085-100G 83-0085-500G
100208-978EA 556.25 USD
100208-978 100208-980
Bismuth ≥99.999%, shot, elongated
Bismuth
Formula: Bi
Boiling Pt: 1560 °C
Melting Pt: 271.3 °C
Density: 9.78 g/cm³ (20 °C)
MDL Number: MFCD00134033
CAS Number: 7440-69-9
EINECS: 231-177-4
Merck Index: 12,01297

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR