Bismuth ≥99.9999%, rods
Supplier: STREM CHEMICALS INC MS
83-0080-25G
83-0080-100G
100196-976EA
510.95
USD
100196-976
100210-916
Bismuth ≥99.9999%, rods
Bismuth
Formula:
Bi Boiling Pt: 1560 °C Melting Pt: 271.3 °C Density: 9.78 g/cm³ (20 °C) |
MDL Number:
MFCD00134033 CAS Number: 7440-69-9 EINECS: 231-177-4 Merck Index: 12,01297 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...