Bismuth ≥99.99%, pieces
Supplier: STREM CHEMICALS INC MS
83-0075-1000G
83-0075-250G
100202-780EA
307.14
USD
100202-780
100210-106
Bismuth ≥99.99%, pieces
Bismuth
Formula:
Bi MW: 208.98 g/mol Boiling Pt: 1560 °C Melting Pt: 271.3 °C Density: 9.80 |
MDL Number:
MFCD00134033 CAS Number: 7440-69-9 EINECS: 231-177-4 Merck Index: 12,01297 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...