Bismuth ≥99.9999%, granules

Supplier: STREM CHEMICALS INC MS

83-0050-250G 83-0050-50G 83-0050-10G
100204-658EA 2563.26 USD
100204-658 100203-336 100208-976
Bismuth ≥99.9999%, granules
Bismuth
Formula: Bi
Boiling Pt: 1560 °C
Melting Pt: 271.3 °C
Density: 9.78 g/cm³ (20 °C)
MDL Number: MFCD00134033
CAS Number: 7440-69-9
EINECS: 231-177-4
Merck Index: 12,01297

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR