Lead ≥99.999%, rods
Supplier: STREM CHEMICALS INC MS
|
Warning
|
82-0100-250G
82-0100-50G
82-0100-1000G
100197-622EA
539.26
USD
100197-622
100203-334
100198-042
Lead ≥99.999%, rods
Lead
Formula:
Pb Boiling Pt: 1620 °C Melting Pt: 327 °C Density: 11.3437 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134050 CAS Number: 7439-92-1 EINECS: 231-100-4 Merck Index: 12,05410 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...