Cyclopentadienylthallium ≥99% (99.9%-Tl), sublimated

Supplier: STREM CHEMICALS INC MS
Danger

81-0305-5G 81-0305-25G 81-0305-1G
100202-776EA 418.95 USD
100202-776 100207-452 100205-210
Cyclopentadienylthallium ≥99% (99.9%-Tl), sublimated
Cyclopentadienylthallium

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₅H₅Tl
Boiling Pt: 90…110 °C (1 mmHg)
Melting Pt: 75°/0.1mm (subl.)
Storage Temperature: Refrigerator
MDL Number: MFCD00134141
CAS Number: 34822-90-7
EINECS: 252-229-2
UN: 1707
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR