Cyclopentadienylthallium ≥95%

Supplier: STREM CHEMICALS INC MS
Danger

81-0300-50G 81-0300-10G
100200-008EA 703.45 USD
100200-008 100206-854
Cyclopentadienylthallium ≥95%
Cyclopentadienylthallium

Metallocenes, Derivatives & Cp Precursors

Formula: C₅H₅Tl
Boiling Pt: 90 - 110 °C (1 -rr)
Melting Pt: 75 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00134141
CAS Number: 34822-90-7
EINECS: 252-229-2
UN: 1707
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR