Sodium tetrabromoaurate(III) hydrate ≥99.9% (Au basis)

Supplier: STREM CHEMICALS INC MS
Warning

79-3505-5G 79-3505-1G
100201-034EA 796.86 USD
100201-034 100205-208
Sodium tetrabromoaurate(III) hydrate ≥99.9% (Au basis)
Sodium tetrabromoaurate(III) hydrate
Formula: NaAuBr₄·xH₂O(x~2)
MW: 539.57 g/mol (anhy)
MDL Number: MFCD00274624
CAS Number: 52495-41-7
EINECS: 257-962-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR