tetra-Chloroauric(III) acid hydrate ≥99.9985% (Au basis) (49% Au), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Gold(III) chloride hydrate, Hydrogen tetrachloroaurate(III) xhydrate, Hydrogen tetrachloroaurate (III) hydrate, Hydrogen tetrachloroaurate hydrate, Chloroauric acid, tetra-Chloroauric acid hydrate

79-2200-1G 79-2200-5G
100206-846EA 389.23 USD
100206-846 100206-848
tetra-Chloroauric(III) acid hydrate ≥99.9985% (Au basis) (49% Au), PURATREM
tetra-Chloroauric(III) acid hydrate
Formula: HAuCl₄·H₂O
MW: 357.8 g/mol
Density: 1.169 g/cm³ (20 °C)
MDL Number: MFCD00149903
CAS Number: 27988-77-8
EINECS: 240-948-4
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR